accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNS

CAS RN 13599-84-3

IUPAC Name: 1,3-benzothiazol-6-ol
Synonyms: 1,3-benzothiazol-6-ol
Molecular Weight (g/mol): 151.18
Molecular Formula: C7H5NOS
InChi Key: ORIIXCOYEOIFSN-UHFFFAOYSA-N
SMILES: OC1=CC=C2N=CSC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results