accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnH3CNCH3SSH3CNCH3SS

CAS RN 136-23-2

IUPAC Name: zinc(2+) bis((dibutylcarbamothioyl)sulfanide)
Synonyms: zinc dibutyldithiocarbamate
Molecular Weight (g/mol): 474.12
Molecular Formula: C18H36N2S4Zn
InChi Key: BOXSVZNGTQTENJ-UHFFFAOYSA-L
SMILES: [Zn++].CCCCN(CCCC)C([S-])=S.CCCCN(CCCC)C([S-])=S
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)