accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOClClCl

CAS RN 136-32-3

IUPAC Name: sodium 2,4,5-trichlorobenzen-1-olate
Synonyms: sodium 2,4,5-trichlorophenolate
Molecular Weight (g/mol): 219.42
Molecular Formula: C6H2Cl3NaO
InChi Key: KWFOMJTYKROHLX-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results