accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOO

CAS RN 136-36-7

IUPAC Name: 3-hydroxyphenyl benzoate
Synonyms: resorcinol monobenzoate
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: GDESWOTWNNGOMW-UHFFFAOYSA-N
SMILES: OC1=CC=CC(OC(=O)C2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results