accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 136-60-7

IUPAC Name: butyl benzoate
Synonyms: butyl benzoate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: XSIFPSYPOVKYCO-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)