accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13603-44-6

IUPAC Name: 2,6-dimethyl-1,4-dihydropyridin-4-one
Synonyms: 2,6-dimethyl-1H-pyridin-4-one
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: PRAFLUMTYHBEHE-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C=C(C)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results