accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3N(R)(S)HO

CAS RN 136030-00-7

IUPAC Name: (1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
Synonyms: (1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
Molecular Weight (g/mol): 150.2
Molecular Formula: C9H12NO
InChi Key: LOPKSXMQWBYUOI-DTWKUNHWSA-O
SMILES: [NH3+][C@H]1[C@@H](O)CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)