accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13610-49-6

IUPAC Name: 2-(2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl)acetate
Synonyms: 2-(2-oxo-1,3-benzoxazol-3-yl)acetate
Molecular Weight (g/mol): 192.15
Molecular Formula: C9H6NO4
InChi Key: PHIUXGVYFVAGTC-UHFFFAOYSA-M
SMILES: [O-]C(=O)CN1C(=O)OC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results