accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13610-59-8

IUPAC Name: 3-(2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl)propanoate
Synonyms: 3-(2-oxo-1,3-benzoxazol-3-yl)propanoate
Molecular Weight (g/mol): 206.18
Molecular Formula: C10H8NO4
InChi Key: BMLRDVCKJXMASM-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCN1C(=O)OC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results