accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHNa

CAS RN 13613-65-5

IUPAC Name: sodium 3-hydroxybutanoate
Synonyms: sodium 3-hydroxybutyrate
Molecular Weight (g/mol): 126.09
Molecular Formula: C4H7NaO3
InChi Key: NBPUSGBJDWCHKC-UHFFFAOYNA-M
SMILES: [Na+].CC(O)CC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results