accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOONa(E/Z)

CAS RN 13615-46-8

IUPAC Name: sodium 3-(1H-indol-3-yl)prop-2-enoate
Synonyms: sodium 3-indoleacrylate
Molecular Weight (g/mol): 209.18
Molecular Formula: C11H8NNaO2
InChi Key: RVUGOFHDOUJAQL-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C=CC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results