accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13623-94-4

IUPAC Name: 1,1-bis(methylsulfanyl)-2-nitroethene
Synonyms: 1,1-bis(methylsulfanyl)-2-nitroethene
Molecular Weight (g/mol): 165.23
Molecular Formula: C4H7NO2S2
InChi Key: NXGHEDHQXXXTTP-UHFFFAOYSA-N
SMILES: CSC(SC)=C[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)