accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 136315-70-3

CH3H3CCH3OOHN(S)(R)OO

CAS RN 136315-70-3

IUPAC Name: (1R,2S)-2-{[(tert-butoxy)carbonyl]amino}cyclopentane-1-carboxylate
Synonyms: (1R,2S)-2-[(tert-butoxycarbonyl)amino]cyclopentane-1-carboxylate
Molecular Weight (g/mol): 228.27
Molecular Formula: C11H18NO4
InChi Key: BUEPEVBYNBQNED-SFYZADRCSA-M
SMILES: CC(C)(C)OC(=O)N[C@H]1CCC[C@H]1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results