accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13637-71-3

IUPAC Name: nickel(2+) diperchlorate
Synonyms: nickel diperchlorate|perchloric acid, nickel(II) salt
Molecular Weight (g/mol): 257.59
Molecular Formula: Cl2NiO8
InChi Key: ZLQBNKOPBDZKDP-UHFFFAOYSA-L
SMILES: [Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results