accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 136370-19-9

IUPAC Name: [4-(heptyloxy)phenyl]boronic acid
Synonyms: 4-(heptyloxy)phenylboronic acid
Molecular Weight (g/mol): 236.12
Molecular Formula: C13H21BO3
InChi Key: UHRONCCLURKEKO-UHFFFAOYSA-N
SMILES: CCCCCCCOC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results