CAS RN 13640-78-3
IUPAC Name:
2-[2-(thiophen-2-yl)ethenyl]thiophene
Synonyms:
2-[2-(thiophen-2-yl)ethenyl]thiophene
Molecular Weight (g/mol):
192.29
Molecular Formula:
C10H8S2
InChi Key:
AYBFWHPZXYPJFW-UHFFFAOYSA-N
SMILES:
S1C=CC=C1C=CC1=CC=CS1
1
–
1
of
1
results
1
trans-1,2-Di(2-thienyl)ethylene 98.0+%, TCI America™
CAS: 13640-78-3 Molecular Formula: C10H8S2 Molecular Weight (g/mol): 192.294 MDL Number: MFCD00145416 InChI Key: AYBFWHPZXYPJFW-AATRIKPKSA-N PubChem CID: 5375272 IUPAC Name: 2-[(E)-2-thiophen-2-ylethenyl]thiophene SMILES: C1=CSC(=C1)C=CC2=CC=CS2