accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3NHO(S)(S)(R)H2N

CAS RN 136465-81-1

IUPAC Name: (3S,4aS,8aR)-3-(tert-butylcarbamoyl)-decahydroisoquinolin-2-ium
Synonyms: (3S,4aS,8aR)-3-(tert-butylcarbamoyl)-decahydroisoquinolin-2-ium
Molecular Weight (g/mol): 239.38
Molecular Formula: C14H27N2O
InChi Key: UPZBXVBPICTBDP-SRVKXCTJSA-O
SMILES: CC(C)(C)NC(=O)[C@@H]1C[C@@H]2CCCC[C@H]2C[NH2+]1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results