accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHNNH2OHO

CAS RN 13650-49-2

IUPAC Name: 2-amino-5-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: 2-amino-5-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 232.28
Molecular Formula: C10H20N2O4
InChi Key: GLZZMUULAVZVTA-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCCCC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)