accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 136507-15-8

IUPAC Name: 2-methoxy-4-nitrobenzaldehyde
Synonyms: 2-methoxy-4-nitrobenzaldehyde
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: LEBUUZXTHMCZQZ-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C=CC(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results