accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13652-09-0

FFFNH2HCl

CAS RN 13652-09-0

IUPAC Name: 2,2,2-trifluoro-1-phenylethan-1-amine hydrochloride
Synonyms: 2,2,2-trifluoro-1-phenylethanamine hydrochloride
Molecular Weight (g/mol): 211.61
Molecular Formula: C8H9ClF3N
InChi Key: LCQGOISHUDYBOS-UHFFFAOYNA-N
SMILES: Cl.NC(C1=CC=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results