CAS RN 136552-06-2
IUPAC Name:
(2S)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}azetidine-2-carboxylate
Synonyms:
(2S)-1-[(9H-fluoren-9-ylmethoxy)carbonyl]azetidine-2-carboxylate
Molecular Weight (g/mol):
322.34
Molecular Formula:
C19H16NO4
InChi Key:
BXRZCDISGRVJCA-KRWDZBQOSA-M
SMILES:
[O-]C(=O)[C@@H]1CCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Fmoc-Azetidine-2-carboxylic ac, Advanced ChemTech
CAS: 136552-06-2 Molecular Formula: C19H16NO4 Molecular Weight (g/mol): 322.34 InChI Key: BXRZCDISGRVJCA-KRWDZBQOSA-M IUPAC Name: (2S)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}azetidine-2-carboxylate SMILES: [O-]C(=O)[C@@H]1CCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12