accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PCH3H3CPH3CCH3CH3CH3OH3COH3COCH3OCH3CH3(?)

CAS RN 1365531-89-0

IUPAC Name: {2'-[bis(3,5-dimethylphenyl)phosphanyl]-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl}bis(3,5-dimethylphenyl)phosphane
Synonyms: {2'-[bis(3,5-dimethylphenyl)phosphanyl]-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl}bis(3,5-dimethylphenyl)phosphane
Molecular Weight (g/mol): 754.89
Molecular Formula: C48H52O4P2
InChi Key: IDNKFBDRZBLTFQ-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C(=C1)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1)C1=C(OC)C=C(OC)C=C1P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results