accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OH3CH3CH3CCH3PH3CH3CCH3OH3CCH3H3CCH3PCH3CH3H3COH3CH3CCH3CH3H3CCH3CH3OH3CCH3H3CCH3OCH3OCH3OCH3OCH3(?)

CAS RN 1365531-98-1

IUPAC Name: {2'-[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphanyl]-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl}bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphane
Synonyms: {2'-[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphanyl]-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl}bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphane
Molecular Weight (g/mol): 1211.64
Molecular Formula: C76H108O8P2
InChi Key: KACYLFSRRUJDSY-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C(=C1)P(C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C1=C(OC)C=C(OC)C=C1P(C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results