accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH(R)OHNOOOFFFFF

CAS RN 136554-94-4

IUPAC Name: 2,3,4,5,6-pentafluorophenyl (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-indol-3-yl)propanoate
Synonyms: 2,3,4,5,6-pentafluorophenyl (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate|FMOC-D-TRP-OPFP
Molecular Weight (g/mol): 592.52
Molecular Formula: C32H21F5N2O4
InChi Key: KLPGTAYCHANVFY-XMMPIXPASA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)[C@@H](CC2=CNC3=CC=CC=C23)NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)