accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13657-16-4

IUPAC Name: 3-benzyl-1,3-oxazolidine
Synonyms: 3-benzyl-1,3-oxazolidine
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: FQNODHUYZYLTPN-UHFFFAOYSA-N
SMILES: C(N1CCOC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results