accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONON

CAS RN 13663-23-5

IUPAC Name: 1-(4-nitrophenyl)azepane
Synonyms: 1-(4-nitrophenyl)azepane
Molecular Weight (g/mol): 220.27
Molecular Formula: C12H16N2O2
InChi Key: WXAAQKMTSQDMII-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N1CCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results