accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13673-62-6

IUPAC Name: 2-(methylsulfanyl)-1,3-benzoxazole
Synonyms: 2-(methylsulfanyl)-1,3-benzoxazole
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: CBXAWZGFEDZKFR-UHFFFAOYSA-N
SMILES: CSC1=NC2=CC=CC=C2O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results