accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13675-18-8

IUPAC Name: (dihydroxyboranyl)boronic acid
Synonyms: tetrahydroxydiborane|hypodiboric acid|diborane(4)tetrol|tetrahydroxydiboron|diboron tetrahydroxide
Molecular Weight (g/mol): 89.65
Molecular Formula: B2H4O4
InChi Key: SKOWZLGOFVSKLB-UHFFFAOYSA-N
SMILES: OB(O)B(O)O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)