accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13676-54-5

IUPAC Name: 1-(4-{[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenyl]methyl}phenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(4-{[4-(2,5-dioxopyrrol-1-yl)phenyl]methyl}phenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 358.35
Molecular Formula: C21H14N2O4
InChi Key: XQUPVDVFXZDTLT-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CC=C(CC2=CC=C(C=C2)N2C(=O)C=CC2=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)