CAS RN 136765-36-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Hydrocodone-D3 Solution, 100 μg/mL in methanol, Sold by MilliporeSigma™ Supelco™
This certified Snap-N-Spike™ solution is suitable for quantitation of hydrocodone levels or isotope dilution methods by LC/MS or GC/MS for pain prescription monitoring, clinical toxicology, pharmaceutical research, or forensic analysis.