accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13679-61-3

CH3SOO

CAS RN 13679-61-3

IUPAC Name: (furan-2-yl)(methylsulfanyl)methanone
Synonyms: furan-2-yl(methylsulfanyl)methanone
Molecular Weight (g/mol): 142.17
Molecular Formula: C6H6O2S
InChi Key: ISKUAGFDTRLBHG-UHFFFAOYSA-N
SMILES: CSC(=O)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results