accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13679-85-1

IUPAC Name: 2-methylthiolan-3-one
Synonyms: 2-methylthiolan-3-one
Molecular Weight (g/mol): 116.18
Molecular Formula: C5H8OS
InChi Key: YMZZPMVKABUEBL-UHFFFAOYNA-N
SMILES: CC1SCCC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results