accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 13686-49-2

IUPAC Name: 1-(2-bromoethyl)naphthalene
Synonyms: 1-(2-bromoethyl)naphthalene
Molecular Weight (g/mol): 235.12
Molecular Formula: C12H11Br
InChi Key: GPHCPUFIWQJZOI-UHFFFAOYSA-N
SMILES: BrCCC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)