CAS RN 13688-55-6
IUPAC Name:
trimethylsilyl prop-2-enoate
Synonyms:
trimethylsilyl prop-2-enoate
Molecular Weight (g/mol):
144.25
Molecular Formula:
C6H12O2Si
InChi Key:
OTYBJBJYBGWBHB-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)OC(=O)C=C
1
–
1
of
1
results
1
Trimethylsilyl Acrylate (stabilized with BHT) 95.0+%, TCI America™
CAS: 13688-55-6 Molecular Formula: C6H12O2Si Molecular Weight (g/mol): 144.245 MDL Number: MFCD00156432 InChI Key: OTYBJBJYBGWBHB-UHFFFAOYSA-N Synonym: Acrylic Acid Trimethylsilyl Ester PubChem CID: 554662 IUPAC Name: trimethylsilyl prop-2-enoate SMILES: C[Si](C)(C)OC(=O)C=C