accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13688-67-0

IUPAC Name: undeca-1,10-diene
Synonyms: undeca-1,10-diene
Molecular Weight (g/mol): 152.28
Molecular Formula: C11H20
InChi Key: VOSLXTGMYNYCPW-UHFFFAOYSA-N
SMILES: C=CCCCCCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results