CAS RN 13688-68-1
IUPAC Name:
8,8-dimethyl-8-silatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene
Synonyms:
8,8-dimethyl-8-silatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene
Molecular Weight (g/mol):
210.35
Molecular Formula:
C14H14Si
InChi Key:
YJQFTKVGILQPMC-UHFFFAOYSA-N
SMILES:
C[Si]1(C)C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
9,9-Dimethyl-9H-9-silafluorene 98.0+%, TCI America™
CAS: 13688-68-1 Molecular Formula: C14H14Si Molecular Weight (g/mol): 210.351 InChI Key: YJQFTKVGILQPMC-UHFFFAOYSA-N Synonym: 5,5-Dimethyl-5H-dibenzo[b,d]silole PubChem CID: 139544 IUPAC Name: 5,5-dimethylbenzo[b][1]benzosilole SMILES: C[Si]1(C2=CC=CC=C2C3=CC=CC=C31)C