accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1369328-62-0

IUPAC Name: {5'-formyl-[2,2'-bithiophen]-5-yl}boronic acid
Synonyms: 5'-formyl-[2,2'-bithiophen]-5-ylboronic acid
Molecular Weight (g/mol): 238.08
Molecular Formula: C9H7BO3S2
InChi Key: RDIOTSOSMWVMSX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(S1)C1=CC=C(S1)C=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results