accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSHOCH3O

CAS RN 136954-20-6

IUPAC Name: 3-sulfanylhexyl acetate
Synonyms: 3-sulfanylhexyl acetate
Molecular Weight (g/mol): 176.27
Molecular Formula: C8H16O2S
InChi Key: JUCARGIKESIVLB-UHFFFAOYNA-N
SMILES: CCCC(S)CCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results