accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 137-07-5

IUPAC Name: 2-aminobenzene-1-thiol
Synonyms: 2-aminothiophenol|O-aminothiophenol
Molecular Weight (g/mol): 125.19
Molecular Formula: C6H7NS
InChi Key: VRVRGVPWCUEOGV-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1S
Boiling Point 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)