accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NH2H3C

CAS RN 137-17-7

IUPAC Name: 2,4,5-trimethylaniline
Synonyms: 2,4,5-trimethylaniline
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: BMIPMKQAAJKBKP-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results