accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CSSCuSSNCH3CH3

CAS RN 137-29-1

IUPAC Name: bis[(dimethylcarbamothioyl)sulfanyl]copper
Synonyms: bis[(dimethylcarbamothioyl)sulfanyl]copper
Molecular Weight (g/mol): 303.96
Molecular Formula: C6H12CuN2S4
InChi Key: ZOUQIAGHKFLHIA-UHFFFAOYSA-L
SMILES: CN(C)C(=S)S[Cu]SC(=S)N(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)