accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCSCH

CAS RN 13702-09-5

IUPAC Name: 3-(prop-2-yn-1-ylsulfanyl)prop-1-yne
Synonyms: 3-(prop-2-yn-1-ylsulfanyl)prop-1-yne
Molecular Weight (g/mol): 110.17
Molecular Formula: C6H6S
InChi Key: XPTMJJIPRSWBDK-UHFFFAOYSA-N
SMILES: C#CCSCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results