accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNHBNHBHNBrBrBr

CAS RN 13703-88-3

IUPAC Name: 2,4,6-tribromo-1,3,5,2,4,6-triazatriborinane
Synonyms: 2,4,6-tribromo-1,3,5,2,4,6-triazatriborinane
Molecular Weight (g/mol): 317.19
Molecular Formula: B3Br3H3N3
InChi Key: PCLMMJIWMGUYKF-UHFFFAOYSA-N
SMILES: BrB1NB(Br)NB(Br)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results