accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 13704-09-1

IUPAC Name: ethyl 2-hydroxy-2-phenylacetate
Synonyms: ethyl mandelate mandelsaeureaethylester ethyl phenylglycolate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: SAXHIDRUJXPDOD-UHFFFAOYNA-N
SMILES: CCOC(=O)C(O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results