accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OCH3O

CAS RN 13706-86-0

IUPAC Name: 5-methylhexane-2,3-dione
Synonyms: 2,3-hexanedione, 5-methyl-
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: PQCLJXVUAWLNSV-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results