accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13709-38-1

IUPAC Name: lanthanum(3+) trifluoride
Synonyms: lanthanum(3+) trifluoride
Molecular Weight (g/mol): 195.9
Molecular Formula: F3La
InChi Key: BYMUNNMMXKDFEZ-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[La+3]
Molecular Weight (g/mol) 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (2)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)