accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSc

CAS RN 13709-47-2

IUPAC Name: scandium(3+) trifluoride
Synonyms: scandium fluoride
Molecular Weight (g/mol): 101.95
Molecular Formula: F3Sc
InChi Key: OEKDNFRQVZLFBZ-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Sc+3]
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results