CAS RN 13709-52-9
IUPAC Name:
tetrahydrogen hafnium tetrafluoride
Synonyms:
hafnium tetrafluoride
Molecular Weight (g/mol):
258.52
Molecular Formula:
F4H4Hf
InChi Key:
HRTPXFIQXNXHQB-UHFFFAOYSA-N
SMILES:
[H+].[H+].[H+].[H+].[F-].[F-].[F-].[F-].[Hf]
1
–
1
of
1
results
1
Hafnium(IV) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 13709-52-9 Molecular Formula: HfF4 MDL Number: MFCD00016127