accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONCl

CAS RN 13720-94-0

IUPAC Name: ethyl 4-chloroquinoline-3-carboxylate
Synonyms: ethyl 4-chloroquinoline-3-carboxylate
Molecular Weight (g/mol): 235.67
Molecular Formula: C12H10ClNO2
InChi Key: DWXQUAHMZWZXHP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C=CC=CC2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results