accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOOH

CAS RN 13721-01-2

IUPAC Name: 4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: 4-oxo-1H-quinoline-3-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: ILNJBIQQAIIMEY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results